Learn More
Medchemexpress LLC HY-13536 5mg Medchemexpress, AZD-2461 CAS:1174043-16-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135365MG
Encompass_Preferred
Medchemexpress, HY-13536 5mg AZD-2461 CAS:1174043-16-3 AZD-2461 is a potent PARP inhibitor, with IC50s of 5 nM, 2 nM and 200 nM for PARP1, PARP2 and PARP3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1384