Learn More
Selleck Chemical LLC AZD4635 (HTL1071) 5mg 1321514-06-0
SDP
Supplier:  Selleck Chemical LLC S87205MG
Encompass_Preferred
AZD4635 (HTL1071) is an oral A2AR antagonist that binds to human A2AR with a Ki of 1.7 nM and with > 30-fold selectivity over other adenosine receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8408