Learn More
Medchemexpress LLC Azd-4818 | 1003566-93-5 | 99.1% | 567.46 g/mol | C27H32Cl2N2O7 | 10 MG
SDP
Catalog No. 5000287124
Encompass_Preferred
AZD-4818 is a potent, orally active antagonist of the chemokine receptor CCR1 used in respiratory inflammation research. It blocks CCR1-mediated recruitment of inflammatory cells and has been evaluated in preclinical models and clinical tolerability studies for chronic obstructive pulmonary disease. Supplied as a solid with documented solubility and storage instructions for laboratory use.
  • Potent CCR1 antagonism, reducing monocyte and neutrophil recruitment.
  • Orally bioavailable profile suitable for dosing studies.
  • High chemical purity for reliable experimental results.
  • Provided with solubility and storage data for reproducible handling.
  • Multiple pack sizes available for flexible experimental needs.
Catalog No. 50-002-87124 Supplier Medchemexpress LLC Supplier No. HY1554510MG
AZD-4818 is a potent, orally active antagonist of the chemokine receptor CCR1 used in respiratory inflammation research. It blocks CCR1-mediated recruitment of inflammatory cells and has been evaluated in preclinical models and clinical tolerability studies for chronic obstructive pulmonary disease. Supplied as a solid with documented solubility and storage instructions for laboratory use.
  • Potent CCR1 antagonism, reducing monocyte and neutrophil recruitment.
  • Orally bioavailable profile suitable for dosing studies.
  • High chemical purity for reliable experimental results.
  • Provided with solubility and storage data for reproducible handling.
  • Multiple pack sizes available for flexible experimental needs.