Learn More
TARGETMOL CHEMICALS INC AZD-4877 5MG
SDP
Catalog No. 502578974
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AZD-4877 is a potent spindle kinesin (Eg5) inhibitor with an IC50 value of 2 nM. AZD-4877 is an alternative configuration of Ispinesib that inhibits mitosis induces the formation of a unipolar spindle phenotype and mediates apoptosis. AZD-4877 has antitumor activity and inhibits circulating peripheral blood mononuclear cells (PBMC). purity: 99%
Catalog No. 50-257-8974 Supplier TARGETMOL CHEMICALS INC Supplier No. T96825MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AZD-4877 is a potent spindle kinesin (Eg5) inhibitor with an IC50 value of 2 nM. AZD-4877 is an alternative configuration of Ispinesib that inhibits mitosis induces the formation of a unipolar spindle phenotype and mediates apoptosis. AZD-4877 has antitumor activity and inhibits circulating peripheral blood mononuclear cells (PBMC). purity: 99%