Learn More
Medchemexpress LLC HY-19855 10mg , AZD-5069 CAS:878385-84-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1985510MG
Encompass_Preferred
HY-19855 10mg AZD-5069 CAS: 878385-84-3 AZD-5069 is a potent CXCR2 chemokine receptor antagonist, used for caner treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1058