Learn More
Abcam AZD-5069, 5MG
Catalog No. 502671239
Encompass_Preferred
MW 476.5 Da, Purity >=98%. A potent and reversible CXCR2 chemokine receptor antagonist with potential anti-inflammatory and antineoplastic activities. Upon administration, CXC chemokine receptor 2 antagonist AZD5069 directly binds to CXCR2 and inhibits its activation. This inhibits CXCR2-mediated signaling and may inhibit tumor cell proliferation in CXCR2-overexpressing tumor cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1239 Supplier Abcam Supplier No. AB2854345MG
MW 476.5 Da, Purity >=98%. A potent and reversible CXCR2 chemokine receptor antagonist with potential anti-inflammatory and antineoplastic activities. Upon administration, CXC chemokine receptor 2 antagonist AZD5069 directly binds to CXCR2 and inhibits its activation. This inhibits CXCR2-mediated signaling and may inhibit tumor cell proliferation in CXCR2-overexpressing tumor cells.
The product is subject to the following: Abcam Restricted Use Statement