Learn More
Selleck Chemical LLC AZD-5462 E1995-5MG
SDP
Catalog No. 5000039235
Encompass_Preferred
AZD-5462(Example 1) is a modulator of RXFP1 the cognate receptor for human relaxin that belongs to the GPCR family 1c number AZD-5462 exhibits anti-fibrotic and anti-inflammatory properties
Catalog No. 50-000-39235 Supplier Selleck Chemical LLC Supplier No. E19955MG
AZD-5462(Example 1) is a modulator of RXFP1 the cognate receptor for human relaxin that belongs to the GPCR family 1c number AZD-5462 exhibits anti-fibrotic and anti-inflammatory properties