Learn More
Medchemexpress LLC AZD-5672 | 780750-65-4 | 95.7% | 632.78 | 5 MG
SDP
Catalog No. 5000364252
Encompass_Preferred
AZD-5672 is an orally active, potent, and selective CCR5 antagonist with an IC50 of 0.32 nM. It demonstrates moderate activity against the hERG ion channel (binding IC50=7.3 μM) and is a substrate of human P-gp, inhibiting P-gp-mediated digoxin transport (IC50=32 μM). It can be used for research related to rheumatoid arthritis.
  • Potent and selective CCR5 antagonist
  • Moderate activity against hERG ion channel
  • Inhibits P-gp-mediated digoxin transport
  • Useful for rheumatoid arthritis research
Catalog No. 50-003-64252 Supplier Medchemexpress LLC Supplier No. 5MGHY119101
AZD-5672 is an orally active, potent, and selective CCR5 antagonist with an IC50 of 0.32 nM. It demonstrates moderate activity against the hERG ion channel (binding IC50=7.3 μM) and is a substrate of human P-gp, inhibiting P-gp-mediated digoxin transport (IC50=32 μM). It can be used for research related to rheumatoid arthritis.
  • Potent and selective CCR5 antagonist
  • Moderate activity against hERG ion channel
  • Inhibits P-gp-mediated digoxin transport
  • Useful for rheumatoid arthritis research