Learn More
Medchemexpress LLC HY-101533 50mg Medchemexpress, AZD-5991 CAS:2143061-81-6 Purity:>98%
SDP
Catalog No. 501932160
Encompass_Preferred
Medchemexpress, HY-101533 50mg AZD-5991 CAS:2143061-81-6 AZD-5991 is a potent and selective Mcl-1 inhibitor with an IC 50 of 0.7 nM in FRET assay and a K d of 0.17 nM in surface plasmon resonance (SPR) assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2160 Supplier Medchemexpress LLC Supplier No. HY10153350MG
Medchemexpress, HY-101533 50mg AZD-5991 CAS:2143061-81-6 AZD-5991 is a potent and selective Mcl-1 inhibitor with an IC 50 of 0.7 nM in FRET assay and a K d of 0.17 nM in surface plasmon resonance (SPR) assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.