Learn More
Medchemexpress LLC HY-19872 1mg Medchemexpress, AZD-6280 CAS:942436-93-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY198721MG
Encompass_Preferred
Medchemexpress, HY-19872 1mg AZD-6280 CAS:942436-93-3 AZD-6280 is a selective GABAA(α2/3) receptor modulator, used for treatment of generalized anxiety disorder. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0041