Learn More
Medchemexpress LLC HY-10344 10mg , AZD 6482 CAS:1173900-33-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1034410MG
Encompass_Preferred
HY-10344 10mg AZD 6482 CAS: 1173900-33-8 AZD 6482 (KIN-193) is a potent and selective p110β inhibitor with an IC50 of 0.69 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9438