Learn More
Medchemexpress LLC AZD 6482 (KIN-193) | 1173900-33-8 | MFCD16659062 | 99.9% | 408.45 g/mol | C22H24N4O4 | 100 MG
SDP
Catalog No. 5000244937
Encompass_Preferred
AZD 6482 is a potent, selective inhibitor of the p110β (PI3Kβ) isoform of phosphoinositide 3-kinase, used as a research tool to probe PI3Kβ-dependent signaling in biochemical and cellular assays. It offers subnanomolar potency against PI3Kβ with a characterized selectivity profile across related PI3K isoforms.
  • Potent PI3Kβ inhibitor with IC50 = 0.69 nM.
  • Selectivity profile includes higher IC50 values for PI3Kδ, PI3Kγ, and PI3Kα.
  • High chemical purity (99.93%).
  • Molecular formula C22H24N4O4; molecular weight 408.45 g/mol.
  • Available as solid and as 10 mM solution in DMSO for assay use.
  • For laboratory research use only; not for human or clinical applications.
Catalog No. 50-002-44937 Supplier Medchemexpress LLC Supplier No. HY10344100MG
AZD 6482 is a potent, selective inhibitor of the p110β (PI3Kβ) isoform of phosphoinositide 3-kinase, used as a research tool to probe PI3Kβ-dependent signaling in biochemical and cellular assays. It offers subnanomolar potency against PI3Kβ with a characterized selectivity profile across related PI3K isoforms.
  • Potent PI3Kβ inhibitor with IC50 = 0.69 nM.
  • Selectivity profile includes higher IC50 values for PI3Kδ, PI3Kγ, and PI3Kα.
  • High chemical purity (99.93%).
  • Molecular formula C22H24N4O4; molecular weight 408.45 g/mol.
  • Available as solid and as 10 mM solution in DMSO for assay use.
  • For laboratory research use only; not for human or clinical applications.