Learn More
AdipoGen AZD 7545
SDP
Catalog No. 502562216
Encompass_Preferred
Chemical. CAS 252017-04-2. Formula C19H18ClF3N2O5S. MW 478.9. . Potent and selective pyruvate dehydrogenase kinase 2 PDK2 inhibitor IC50=6.4nM, consequently increasing pyruvate dehydrogenase PDH activity. Less potent inhibitor towards PDK1 and PDK3 IC50=87 and 600nM, respectively. Inhibitor of glycolysis. Antiobesity and antidiabetic agent. Improves blood glucose control in obese fa/fa Zucker rats. Useful agent for immunometabolism research. To maintain a continuous and steady supply of ATP during the feed-fast cycle, cells must select fatty acid or glucose for fuel. This process is largely controlled by the pyruvate dehydrogenase complex PDC, which regulates the entry of glycolytic products into the tricarboxylic acid cycle by catalyzing the oxidative decarboxylation of pyruvate to acetyl-coenzyme A CoA in the mitochondria of mammalian cells.
Catalog No. 50-256-2216 Supplier AdipoGen Supplier No. AGCR13692M001
Chemical. CAS 252017-04-2. Formula C19H18ClF3N2O5S. MW 478.9. . Potent and selective pyruvate dehydrogenase kinase 2 PDK2 inhibitor IC50=6.4nM, consequently increasing pyruvate dehydrogenase PDH activity. Less potent inhibitor towards PDK1 and PDK3 IC50=87 and 600nM, respectively. Inhibitor of glycolysis. Antiobesity and antidiabetic agent. Improves blood glucose control in obese fa/fa Zucker rats. Useful agent for immunometabolism research. To maintain a continuous and steady supply of ATP during the feed-fast cycle, cells must select fatty acid or glucose for fuel. This process is largely controlled by the pyruvate dehydrogenase complex PDC, which regulates the entry of glycolytic products into the tricarboxylic acid cycle by catalyzing the oxidative decarboxylation of pyruvate to acetyl-coenzyme A CoA in the mitochondria of mammalian cells.