Learn More
Medchemexpress LLC HY-111783 25mg , AZD-7648 CAS:2230820-11-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11178325MG
Encompass_Preferred
HY-111783 25mg AZD-7648 CAS: 2230820-11-6 AZD-7648 is a potent, orally active, selective DNA-PK inhibitor with an IC50 of 0.6 nM. AZD-7648 induces apoptosis and shows antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0535