Learn More
Medchemexpress LLC HY-10992 10mg Medchemexpress, AZD-7762 CAS:860352-01-8 Purity:>98%
SDP
Catalog No. 502028801
Encompass_Preferred
Medchemexpress, HY-10992 10mg AZD-7762 CAS:860352-01-8 AZD-7762 is a potent ATP-competitive checkpoint kinase (Chk) inhibitor in with an IC50 of 5 nM for Chk1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8801 Supplier Medchemexpress LLC Supplier No. HY1099210MG
Medchemexpress, HY-10992 10mg AZD-7762 CAS:860352-01-8 AZD-7762 is a potent ATP-competitive checkpoint kinase (Chk) inhibitor in with an IC50 of 5 nM for Chk1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.