Learn More
Medchemexpress LLC HY-10992 100mg , AZD-7762 CAS:860352-01-8 Purity:98%
SDP
Catalog No. 501492773
Encompass_Preferred
Medchemexpress, HY-10992 100mg AZD-7762 CAS:860352-01-8 Formula:C17H19FN4O2S IC50: 5 nM (ChK1), 5 nM (ChK2) Purity:98% AZD-7762 is a potent ATP-competitive checkpoint kinase ( Chk ) inhibitor in with IC 50 of 5 nM for Chk1. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2773 Supplier Medchemexpress LLC Supplier No. HY10992100MG
Medchemexpress, HY-10992 100mg AZD-7762 CAS:860352-01-8 Formula:C17H19FN4O2S IC50: 5 nM (ChK1), 5 nM (ChK2) Purity:98% AZD-7762 is a potent ATP-competitive checkpoint kinase ( Chk ) inhibitor in with IC 50 of 5 nM for Chk1. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.