Learn More
Medchemexpress LLC AZD-7762 hydrochloride | 1246094-78-9 | 98.03% | 398.88 | 1 MG
SDP
Catalog No. 5000345082
Encompass_Preferred
AZD-7762 hydrochloride is a potent ATP-competitive checkpoint kinase (Chk) inhibitor. It inhibits Chk1 and Chk2, abrogates DNA damage-induced S and G2 checkpoints, and enhances the efficacy of DNA-targeted therapies. This compound modulates downstream checkpoint pathway proteins and has been studied in clinical trials for various cancers.
  • Potently inhibits Chk1 and Chk2
  • Abrogates DNA damage-induced S and G2 checkpoints
  • Enhances efficacy of certain DNA-targeted therapies
  • Modulates downstream checkpoint pathway proteins
  • Binds to the ATP-binding site of Chk1 in a reversible manner
Catalog No. 50-003-45082 Supplier Medchemexpress LLC Supplier No. HY10992A50MG
AZD-7762 hydrochloride is a potent ATP-competitive checkpoint kinase (Chk) inhibitor. It inhibits Chk1 and Chk2, abrogates DNA damage-induced S and G2 checkpoints, and enhances the efficacy of DNA-targeted therapies. This compound modulates downstream checkpoint pathway proteins and has been studied in clinical trials for various cancers.
  • Potently inhibits Chk1 and Chk2
  • Abrogates DNA damage-induced S and G2 checkpoints
  • Enhances efficacy of certain DNA-targeted therapies
  • Modulates downstream checkpoint pathway proteins
  • Binds to the ATP-binding site of Chk1 in a reversible manner