Learn More
Medchemexpress LLC Azd-7762 hydrochloride | 1246094-78-9 | 99.58% | 398.88 | 25 MG
SDP
Numéro de catalogue. 5000347572
Encompass_Preferred
AZD-7762 hydrochloride is a potent ATP-competitive checkpoint kinase (Chk) inhibitor with an IC50 of 5 nM for Chk1. It inhibits Chk1 and Chk2, abrogates DNA damage-induced S and G2 checkpoints, enhances the efficacy of NSC 613327 and SKF 104864A, and modulates downstream checkpoint pathway proteins. It potentiates antitumor activity and induces lymphoma cell death in combination with CX-5461.
  • Potent ATP-competitive checkpoint kinase (Chk) inhibitor.
  • IC50 of 5 nM for Chk1.
  • Equally potent inhibitor of Chk1 and Chk2 in vitro.
  • Abrogates DNA damage-induced S and G2 checkpoints.
  • Enhances the efficacy of NSC 613327 and SKF 104864A.
  • Modulates downstream checkpoint pathway proteins.
  • Binds to the ATP-binding site of Chk1.
  • Potentiates antitumor activity in xenograft studies.
  • Induces lymphoma cell death in combination with CX-5461.
Numéro de catalogue. 50-003-47572 Fournisseur Medchemexpress LLC Code fournisseur 25MGHY10992A
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
AZD-7762 hydrochloride is a potent ATP-competitive checkpoint kinase (Chk) inhibitor with an IC50 of 5 nM for Chk1. It inhibits Chk1 and Chk2, abrogates DNA damage-induced S and G2 checkpoints, enhances the efficacy of NSC 613327 and SKF 104864A, and modulates downstream checkpoint pathway proteins. It potentiates antitumor activity and induces lymphoma cell death in combination with CX-5461.
  • Potent ATP-competitive checkpoint kinase (Chk) inhibitor.
  • IC50 of 5 nM for Chk1.
  • Equally potent inhibitor of Chk1 and Chk2 in vitro.
  • Abrogates DNA damage-induced S and G2 checkpoints.
  • Enhances the efficacy of NSC 613327 and SKF 104864A.
  • Modulates downstream checkpoint pathway proteins.
  • Binds to the ATP-binding site of Chk1.
  • Potentiates antitumor activity in xenograft studies.
  • Induces lymphoma cell death in combination with CX-5461.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.