Learn More
AdipoGen AZD-8055, 1009298-09-2, MFCD16660191, 1 mg
SDP
Supplier:  AdipoGen SYN1166M001
Encompass_Preferred
MF: C25H31N5O4, MW: 465.5, ≥95%, Appearance: Solid. AZD8055 is a novel ATP-competitive inhibitor of mTOR kinase activity with an IC(50) of 0.8nM. It shows excellent selectivity (~1,000-fold) against all class I phosphatidylinositol 3-kinase (PI3K) isoforms and other members of the PI3K-like kinase family.
Catalog No. 50-101-5840