Learn More
Medchemexpress LLC Azd-8529 | 1092453-15-0 | MFCD31657360 | 98.0% | 487.47 g/mol | C24H24F3N5O3 | 100 MG
SDP
Catalog No. 5000241037
Encompass_Preferred
A small-molecule positive allosteric modulator of metabotropic glutamate receptor 2 (mGluR2) used in preclinical and clinical research to study receptor modulation, CNS penetration, and behavioral pharmacology. The compound is characterized by established chemical identifiers, molecular weight, and reported potency, and is supplied at high purity for laboratory use.
  • Positive allosteric modulator of mGluR2 with reported nanomolar potency.
  • High purity suitable for in vitro and in vivo research.
  • CNS penetrant with documented pharmacokinetic and safety data.
  • Available in multiple pack sizes and as a ready-to-use DMSO solution.
  • Well-characterized chemical identifiers and spectral data for confirmation.
Catalog No. 50-002-41037 Supplier Medchemexpress LLC Supplier No. HY107457100MG
A small-molecule positive allosteric modulator of metabotropic glutamate receptor 2 (mGluR2) used in preclinical and clinical research to study receptor modulation, CNS penetration, and behavioral pharmacology. The compound is characterized by established chemical identifiers, molecular weight, and reported potency, and is supplied at high purity for laboratory use.
  • Positive allosteric modulator of mGluR2 with reported nanomolar potency.
  • High purity suitable for in vitro and in vivo research.
  • CNS penetrant with documented pharmacokinetic and safety data.
  • Available in multiple pack sizes and as a ready-to-use DMSO solution.
  • Well-characterized chemical identifiers and spectral data for confirmation.