Learn More
Medchemexpress LLC Azd-8529 | 1092453-15-0 | MFCD31657360 | 98.0% | 487.47 g/mol | C24H24F3N5O3 | 5 MG
SDP
Catalog No. 5000239965
Encompass_Preferred
AZD-8529 is a potent, selective, orally bioavailable positive allosteric modulator of the metabotropic glutamate receptor 2 (mGluR2) used in preclinical neuroscience and pharmacology research. The compound is provided with defined purity and physicochemical characterization to support reproducible in vitro and in vivo studies.
  • Potent mGluR2 positive allosteric modulator.
  • High selectivity for mGluR2 over related receptors.
  • Orally bioavailable, suitable for in vivo studies.
  • Provided with defined purity and molecular data for reproducibility.
  • Available in small-quantity formats and DMSO solution for screening.
Catalog No. 50-002-39965 Supplier Medchemexpress LLC Supplier No. HY1074575MG
AZD-8529 is a potent, selective, orally bioavailable positive allosteric modulator of the metabotropic glutamate receptor 2 (mGluR2) used in preclinical neuroscience and pharmacology research. The compound is provided with defined purity and physicochemical characterization to support reproducible in vitro and in vivo studies.
  • Potent mGluR2 positive allosteric modulator.
  • High selectivity for mGluR2 over related receptors.
  • Orally bioavailable, suitable for in vivo studies.
  • Provided with defined purity and molecular data for reproducibility.
  • Available in small-quantity formats and DMSO solution for screening.