Learn More
Medchemexpress LLC Azd-8529 mesylate | 1314217-69-0 | MFCD32645412 | 99.6% | 583.58 g/mol | C25H28F3N5O6S | 5 MG
SDP
Catalog No. 5000239799
Encompass_Preferred
AZD-8529 mesylate is a research-grade positive allosteric modulator of the metabotropic glutamate receptor 2 (mGluR2). Supplied as a characterized, high-purity small-molecule sample, it is intended for in vitro and in vivo pharmacology studies and assay development.
Catalog No. 50-002-39799 Supplier Medchemexpress LLC Supplier No. HY107457A5MG
AZD-8529 mesylate is a research-grade positive allosteric modulator of the metabotropic glutamate receptor 2 (mGluR2). Supplied as a characterized, high-purity small-molecule sample, it is intended for in vitro and in vivo pharmacology studies and assay development.