Learn More
Medchemexpress LLC HY-12869 5mg Medchemexpress, AZD-8835 CAS:1620576-64-8 Purity:>98%
SDP
Catalog No. 502081173
Encompass_Preferred
Medchemexpress, HY-12869 5mg AZD-8835 CAS:1620576-64-8 AZD8835 is a potent and selective inhibitor of PI3Kα and PI3Kδ with IC50s of 6.2 and 5.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1173 Supplier Medchemexpress LLC Supplier No. HY128695MG
Medchemexpress, HY-12869 5mg AZD-8835 CAS:1620576-64-8 AZD8835 is a potent and selective inhibitor of PI3Kα and PI3Kδ with IC50s of 6.2 and 5.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.