Learn More
Apexbio Technology LLC AZD-9291, 10mg. Cas: 1421373-65-0 MFCD: MFCD27988062. Mutated forms EGFR inhibitor
SDP
Supplier:  Apexbio Technology LLC B110410
Encompass_Preferred
AZD-9291 is an irreversible inhibitor of mutant EGFR with IC50 values of 15nM, 17nM and 480nM, respectively for L858R/T790M, ex19del and wild-type EGFR [1].A series of mutations cause the resistance of EGFR, AZD-9291 is developed for an irreversible and selective inhibitor of the mutant EGFR. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5204