Learn More
Medchemexpress LLC HY-15772 500mg , AZD-9291 Osimertinib;Mereletinib CAS:1421373-65-0 Purity:98%
SDP
Catalog No. 501492323
Encompass_Preferred
Medchemexpress, HY-15772 500mg AZD-9291 Osimertinib;Mereletinib CAS:1421373-65-0 Formula:C28H33N7O2 IC50: 1 nM (EGFR L858R/T790M ), 12 nM (EGFR L858R ) Purity:98% AZD-9291 is an irreversible and mutant selective EGFR inhibitor with IC 50 s of 12 and 1 nM against EGFR L858R and EGFR L858R/T790M , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2323 Supplier Medchemexpress LLC Supplier No. HY15772500MG
Add to Cart
Add to Cart
Medchemexpress, HY-15772 500mg AZD-9291 Osimertinib;Mereletinib CAS:1421373-65-0 Formula:C28H33N7O2 IC50: 1 nM (EGFR L858R/T790M ), 12 nM (EGFR L858R ) Purity:98% AZD-9291 is an irreversible and mutant selective EGFR inhibitor with IC 50 s of 12 and 1 nM against EGFR L858R and EGFR L858R/T790M , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.