Learn More
Selleck Chemical LLC AZD-9574-5mg
SDP
Supplier:  Selleck Chemical LLC E21475MG
Encompass_Preferred
AZD-9574 is a novel PARP inhibitor which combines PARP1 selectivity, trapping and high CNS penetration in a single molecule.
Catalog No. 50-239-0055