Learn More
Medchemexpress LLC HY-145804 5mg , AZD-9574 CAS:2756333-39-6 Purity:>98%
SDP
Catalog No. 502261477
Encompass_Preferred
HY-145804 5mg AZD-9574 CAS: 2756333-39-6 AZD-9574 is a potent and brain penetrant PARP1 inhibitor and shows >8000-fold selectivity for PARP1 compared to PARP2/3/5a/6. AZD-9574 acts by selectively inhibiting and trapping PARP1 at the sites of SSBs. AZD-9574 is an anti-cancer agent and can be used for HRD+ breast cancer and advanced solid malignancies research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1477 Supplier Medchemexpress LLC Supplier No. HY1458045MG
HY-145804 5mg AZD-9574 CAS: 2756333-39-6 AZD-9574 is a potent and brain penetrant PARP1 inhibitor and shows >8000-fold selectivity for PARP1 compared to PARP2/3/5a/6. AZD-9574 acts by selectively inhibiting and trapping PARP1 at the sites of SSBs. AZD-9574 is an anti-cancer agent and can be used for HRD+ breast cancer and advanced solid malignancies research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.