Learn More
Medchemexpress LLC HY-136255 5mg Medchemexpress, AZD-9833 CAS:2222844-89-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1362555MG
Encompass_Preferred
Medchemexpress, HY-136255 5mg AZD-9833 CAS:2222844-89-3 AZD-9833 is a potent and orally active estrogen receptor (ER) antagonist. AZD-9833 is used for the study of ER+ HER2-advanced breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0410