Learn More
Medchemexpress LLC HY-100016 10mg , AZD0156 CAS:1821428-35-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10001610MG
Encompass_Preferred
HY-100016 10mg AZD0156 CAS: 1821428-35-6 AZD0156 is a potent, selective and orally active ATM inhibitor with an IC50 of 0.58 nM. AZD0156 inhibits the ATM-mediated signaling, prevents DNA damage checkpoint activation, disrupts DNA damage repair, and induces tumor cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9849