Learn More
Medchemexpress LLC HY-100016 100mg , AZD0156 CAS:1821428-35-6 Purity:98%
Supplier: Medchemexpress LLC HY100016100MG
Medchemexpress, HY-100016 100mg AZD0156 CAS:1821428-35-6 Formula:C26H31N5O3 Purity:98% AZD0156 is an orally active, potent and selective ATM kinase inhibitor, used for cancer research. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.