Learn More
Medchemexpress LLC HY-10127 10mg Medchemexpress, AZD1152 CAS:722543-31-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1012710MG
Encompass_Preferred
Medchemexpress, HY-10127 10mg AZD1152 CAS:722543-31-9 AZD1152 is a pro-drug of Barasertib-hQPA, which is a highly selective Aurora B inhibitor with IC50 of 0.37 nM in a cell-free assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8654