Learn More
Abcam AZD1152-HQPA, Aurora B kinase inhibitor, 5MG
Catalog No. 502706367
Encompass_Preferred
MW 507.6 Da, Purity >99%. Potent, selective Aurora B kinase inhibitor (IC₅₀ = 0.37 nM). Induces apoptosis in vitro. Antitumor agent. Cell-permeable. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6367 Supplier Abcam Supplier No. AB1420495MG
MW 507.6 Da, Purity >99%. Potent, selective Aurora B kinase inhibitor (IC₅₀ = 0.37 nM). Induces apoptosis in vitro. Antitumor agent. Cell-permeable. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement