Learn More
Selleck Chemical LLC AZD1208-10mg
SDP
Supplier:  Selleck Chemical LLC S710410MG
Encompass_Preferred
AZD1208 is a potent, and orally available Pim kinase inhibitor with IC50 of 0.4 nM, 5 nM, and 1.9 nM for Pim1, Pim2, and Pim3 in cell-free assays, respectively. AZD1208 induces autophagy, cell cycle arrest and apoptosis. Phase 1.
Catalog No. 50-238-9652