Learn More
Selleck Chemical LLC AZD1390 5mg 2089288-03-7
SDP
Supplier:  Selleck Chemical LLC S86805MG
Encompass_Preferred
AZD1390 is a first-in-class orally available and CNS penetrant ATM inhibitor with an IC50 of 0.78 nM in cells and >10,000-fold selectivity over closely related members of the PIKK family of enzymes and excellent selectivity across a broad panel of kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1728524