Learn More
Abcam AZD1480, 10MG
Catalog No. 502682941
Encompass_Preferred
MW 348.76 Da, Purity >98%. Novel, potent and selective small-molecule JAK2 inhibitor (IC₅₀ < 3 nM). It can effectively inhibit tumor angiogenesis and metastasis mediated by STAT3 in stromal cells as well as tumor cells. Also demonstrates >50-fold selectivity for JAK2 over JAK3.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2941 Supplier Abcam Supplier No. AB27352610MG
MW 348.76 Da, Purity >98%. Novel, potent and selective small-molecule JAK2 inhibitor (IC₅₀ < 3 nM). It can effectively inhibit tumor angiogenesis and metastasis mediated by STAT3 in stromal cells as well as tumor cells. Also demonstrates >50-fold selectivity for JAK2 over JAK3.
The product is subject to the following: Abcam Restricted Use Statement