Learn More
Selleck Chemical LLC AZD1981 5mg 802904-66-1
SDP
Supplier:  Selleck Chemical LLC S72635MG
Encompass_Preferred
AZD1981 is a potent, selective CRTh2 (DP2) receptor antagonist with IC50 of 4 nM, showing >1000-fold selectivity over more than 340 other enzymes and receptors, including DP1. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4573