Learn More
Apexbio Technology LLC AZD2014, 50mg. Cas: 1009298-59-2 MFCD: MFCD22628784. Novel mTOR inhibitor
SDP
Supplier:  Apexbio Technology LLC A837350
Encompass_Preferred
AZD2014 is a novel, potent and highly selective dual inhibitor of the mammalian rapamycin (mTORC1 and mTORC2) with an IC50 value of 2.8 nM. It is an oral inhibitor and possesses potential antineoplastic activity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5133