Learn More
Selleck Chemical LLC Vistusertib (AZD2014) 5mg 1009298-59-2
SDP
Supplier:  Selleck Chemical LLC S27835MG
Encompass_Preferred
Vistusertib (AZD2014) is a novel mTOR inhibitor with IC50 of 2.8 nM in a cell-free assay; highly selective against multiple PI3K isoforms (?/?/?/?). AZD2014 showed no or weak binding to the majority of kinases when tested at 1 ?M. AZD2014 induces proliferation suppression, apoptosis, cell cycle arrest, and autophagy in HCC cells with antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0379151