Learn More
Medchemexpress LLC AZD2098 | 566203-88-1 | 334.18 | 100 MG
SDP
Catalog No. 5000448537
Encompass_Preferred
AZD2098 is a potent and selective CC-chemokine receptor 4 (CCR4) inhibitor with pIC50s of 7.8, 8.0, 8.0, and 7.6 for human, rat, mouse, and dog, respectively. It is used for asthma research.
  • Potent and selective CCR4 inhibitor
  • Used for asthma research
  • Exhibits efficacy against antigen-induced inflammatory response
  • Purity of 99.91%
Catalog No. 50-004-48537 Supplier Medchemexpress LLC Supplier No. HYU00064100MG
AZD2098 is a potent and selective CC-chemokine receptor 4 (CCR4) inhibitor with pIC50s of 7.8, 8.0, 8.0, and 7.6 for human, rat, mouse, and dog, respectively. It is used for asthma research.
  • Potent and selective CCR4 inhibitor
  • Used for asthma research
  • Exhibits efficacy against antigen-induced inflammatory response
  • Purity of 99.91%