Learn More
Medchemexpress LLC AZD2098 | 566203-88-1 | 334.18 | 50 MG
SDP
Catalog No. 5000447243
Encompass_Preferred
AZD2098 is a potent and selective CC-chemokine receptor 4 (CCR4) inhibitor used for asthma research. It demonstrates pIC50s of 7.8, 8.0, 8.0, and 7.6 for human, rat, mouse, and dog respectively, showing efficacy against antigen-induced inflammatory responses.
  • Potent and selective CC-chemokine receptor 4 (CCR4) inhibitor
  • Suitable for asthma research
  • High purity of 99.91%
  • Soluble in DMSO and ethanol for in vitro studies
  • Comprehensive documentation available, including Data Sheet, SDS, and COA
Catalog No. 50-004-47243 Supplier Medchemexpress LLC Supplier No. HYU0006450MG
AZD2098 is a potent and selective CC-chemokine receptor 4 (CCR4) inhibitor used for asthma research. It demonstrates pIC50s of 7.8, 8.0, 8.0, and 7.6 for human, rat, mouse, and dog respectively, showing efficacy against antigen-induced inflammatory responses.
  • Potent and selective CC-chemokine receptor 4 (CCR4) inhibitor
  • Suitable for asthma research
  • High purity of 99.91%
  • Soluble in DMSO and ethanol for in vitro studies
  • Comprehensive documentation available, including Data Sheet, SDS, and COA