Learn More
Medchemexpress LLC HY-U00064 5mg Medchemexpress, AZD2098 CAS:566203-88-1 Purity:>98%
SDP
Catalog No. 501970619
Encompass_Preferred
Medchemexpress, HY-U00064 5mg AZD2098 CAS:566203-88-1 AZD2098 is a potent and selective CC-chemokine receptor 4 (CCR4) inhibitor with pIC50s of 7.8, 8.0, 8.0 and 7.6 for human, rat, mouse and dog respectively, used for asthma research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0619 Supplier Medchemexpress LLC Supplier No. HYU000645MG
Medchemexpress, HY-U00064 5mg AZD2098 CAS:566203-88-1 AZD2098 is a potent and selective CC-chemokine receptor 4 (CCR4) inhibitor with pIC50s of 7.8, 8.0, 8.0 and 7.6 for human, rat, mouse and dog respectively, used for asthma research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.