Learn More
Medchemexpress LLC HY-135891 5mg Medchemexpress, AZD2423 CAS:1229603-37-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1358915MG
Encompass_Preferred
Medchemexpress, HY-135891 5mg AZD2423 CAS:1229603-37-5 AZD2423 is a potent, selective, orally bioavailable, and non-competitive CCR2 chemokine receptor negative allosteric modulator. AZD2423 has an IC50 of 1.2 nM for CCR2 Ca2+ flux . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1413