Learn More
Apexbio Technology LLC AZD2461 1174043-16-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061545
Encompass_Preferred
AZD2461 (CAS 1174043-16-3) is a novel small-molecule inhibitor of poly(ADP-ribose) polymerase (PARP) exhibiting potency with an IC50 of 5 nM PARP enzymes are pivotal regulators of DNA repair and apoptosis-associated processes In breast cancer cell lines such as SKBR-3 and MCF-7 AZD2461 reduces cell viability in a concentration- and time-dependent manner arresting cells predominantly in the G2 phase In BRCA1-deficient mouse KB1P tumors AZD2461 disrupts PARP activity regardless of P-glycoprotein-mediated resistance to other inhibitors like olaparib highlighting its utility in overcoming drug resistance and elucidating DNA repair pathways in cancer research
Catalog No. 50-000-61545 Supplier Apexbio Technology LLC Supplier No. A41645.1
 Restricted Item
Add to Cart
Add to Cart
AZD2461 (CAS 1174043-16-3) is a novel small-molecule inhibitor of poly(ADP-ribose) polymerase (PARP) exhibiting potency with an IC50 of 5 nM PARP enzymes are pivotal regulators of DNA repair and apoptosis-associated processes In breast cancer cell lines such as SKBR-3 and MCF-7 AZD2461 reduces cell viability in a concentration- and time-dependent manner arresting cells predominantly in the G2 phase In BRCA1-deficient mouse KB1P tumors AZD2461 disrupts PARP activity regardless of P-glycoprotein-mediated resistance to other inhibitors like olaparib highlighting its utility in overcoming drug resistance and elucidating DNA repair pathways in cancer research