Learn More
Selleck Chemical LLC AZD2461 5mg 1174043-16-3
SDP
Catalog No. 501364331
Encompass_Preferred
AZD2461 is a novel PARP inhibitor with low affinity for Pgp than Olaparib. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4331 Supplier Selleck Chemical LLC Supplier No. S70295MG
AZD2461 is a novel PARP inhibitor with low affinity for Pgp than Olaparib. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.