Learn More
Apexbio Technology LLC AZD2461, 5mg. Cas: 1174043-16-3 MFCD: MFCD24386811. Novel PARP inhibitor
SDP
Supplier:  Apexbio Technology LLC A41645
Encompass_Preferred
AZD2461 is a novel PARP inhibitor with IC50 value of 5nM [1].Poly (ADP-ribose) polymerase (PARP) is a family of proteins involved in a number of cellular processes involving mainly DNA repair and programmed cell death [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4897