Learn More
Medchemexpress LLC HY-112802 10mg Medchemexpress, AZD3229 CAS:2248003-60-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11280210MG
Encompass_Preferred
Medchemexpress, HY-112802 10mg AZD3229 CAS:2248003-60-1 AZD3229 is a potent pan-KIT mutant inhibitor for the treatment of gastrointestinal stromal tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9174