Learn More
Medchemexpress LLC HY-112802A 10mg Medchemexpress, AZD3229 Tosylate CAS:2248003-71-4 Purity:>98%
SDP
Catalog No. 501969175
Encompass_Preferred
Medchemexpress, HY-112802A 10mg AZD3229 Tosylate CAS:2248003-71-4 AZD3229 Tosylate is a potent pan-KIT mutant inhibitor for the treatment of gastrointestinal stromal tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9175 Supplier Medchemexpress LLC Supplier No. HY112802A10MG
Medchemexpress, HY-112802A 10mg AZD3229 Tosylate CAS:2248003-71-4 AZD3229 Tosylate is a potent pan-KIT mutant inhibitor for the treatment of gastrointestinal stromal tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.