Learn More
Selleck Chemical LLC Lanabecestat (AZD3293) 5mg 1383982-64-6 LY3314814
SDP
Catalog No. 501365780
Encompass_Preferred
Lanabecestat (AZD3293, LY3314814) is an oral beta-secretase 1 cleaving enzyme (BACE) inhibitor with an inhibitory constant Ki? of 0.4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5780 Supplier Selleck Chemical LLC Supplier No. S81935MG
Lanabecestat (AZD3293, LY3314814) is an oral beta-secretase 1 cleaving enzyme (BACE) inhibitor with an inhibitory constant Ki? of 0.4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.