Learn More
Selleck Chemical LLC AZD3463 5mg 1356962-20-3
SDP
Supplier:  Selleck Chemical LLC S71065MG
Encompass_Preferred
AZD3463 is a novel orally bioavailable ALK inhibitor with Ki of 0.75 nM, which also inhibits IGF1R with equivalent potency. AZD3463 suppresses cell viability by inducing both cell apoptosis and autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4413