Learn More
Selleck Chemical LLC AZD3839 10mg 1227163-84-9
SDP
Supplier:  Selleck Chemical LLC S773110MG
Encompass_Preferred
AZD3839 is a potent and selective BACE1 inhibitor with Ki of 26.1 nM, about 14-fold selectivity over BACE2. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1074336